Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009654.1 | NP_001009654.1 | 2 - 3 | 205 | 91 | |
XM_006234288.2 | XP_006234350.1 | 2 - 3 | 211 | 91 | ||
GenBank mRNA | BC088213.1 | - | 2 - 3 | 205 | 7383479 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |