Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106513.1 | NP_001099983.1 | 3 - 4 | 891 | 84 | |
XM_008762162.1 | XP_008760384.1 | 3 - 4 | 836 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |