Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106498.1 | NP_001099968.1 | 5 - 6 | 1493 | 77 | |
XM_008762091.1 | XP_008760313.1 | 4 - 5 | 2793 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Recommended Assay Rattus norvegicus Probe spans exons |