Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031045.2 | NP_112307.2 | 4 - 5 | 1044 | 98 | |
GenBank mRNA | M29787.1 | - | 4 - 5 | 1044 | 98 | |
X56917.1 | - | 4 - 5 | 1035 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |