Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008353.2 | NP_001008354.1 | 4 - 5 | 1919 | 111 | |
XM_008762254.1 | XP_008760476.1 | 4 - 5 | 1879 | 111 | ||
GenBank mRNA | BC083863.1 | - | 4 - 5 | 1897 | 7127425 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |