Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033927.1 | NP_001029099.1 | 3 - 4 | 318 | 73 | |
GenBank mRNA | BC127457.1 | - | 3 - 4 | 308 | 8363318 | 73 |
CB608471.1 | - | 3 - 4 | 325 | 73 | ||
CK478110.1 | - | 3 - 4 | 318 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |