Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106510.1 | NP_001099980.1 | 12 - 13 | 1808 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |