Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011957.1 | NP_001011957.1 | 6 - 7 | 666 | 82 | |
XM_006234831.2 | XP_006234893.1 | 6 - 7 | 698 | 82 | ||
XM_006234832.2 | XP_006234894.1 | 6 - 7 | 553 | 82 | ||
GenBank mRNA | BC084693.1 | - | 6 - 7 | 666 | 7107022 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |