Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029918.1 | NP_001025089.1 | 1 | 2089 | 170 | |
GenBank mRNA | BC098051.1 | - | 1 | 2089 | 7457462 | 170 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |