Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234874.2 | XP_006234936.1 | 5 - 6 | 1220 | 76 | |
XM_006234875.2 | XP_006234937.1 | 4 - 5 | 1127 | 76 | ||
XM_006234876.2 | XP_006234938.1 | 4 - 5 | 665 | 76 | ||
XM_006234877.1 | XP_006234939.1 | 4 - 5 | 658 | 76 | ||
XM_230512.7 | XP_230512.4 | 4 - 5 | 696 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |