Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Knstrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004264.1 | NP_001004264.1 | 1 - 2 | 279 | 101 | |
XM_006234757.2 | XP_006234819.1 | 1 - 2 | 315 | 101 | ||
GenBank mRNA | BC079438.1 | - | 1 - 2 | 279 | 7115344 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |