Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014180.1 | NP_001014202.1 | 6 - 7 | 494 | 70 | |
XM_006234585.2 | XP_006234647.1 | 6 - 7 | 572 | 70 | ||
GenBank mRNA | BC083756.1 | - | 6 - 7 | 494 | 7192721 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |