Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serping1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199093.1 | NP_954524.1 | 1 - 2 | 103 | 79 | |
XM_006234447.2 | XP_006234509.1 | 2 - 3 | 178 | 79 | ||
GenBank mRNA | BC061860.1 | - | 1 - 2 | 103 | 5598525 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |