Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serping1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199093.1 | NP_954524.1 | 2 - 3 | 622 | 87 | |
XM_006234447.2 | XP_006234509.1 | 3 - 4 | 697 | 87 | ||
GenBank mRNA | BC061860.1 | - | 2 - 3 | 622 | 5598525 | 87 |
FQ219227.1 | - | 1 - 2 | 326 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |