Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031119.2 | NP_112381.2 | 5 - 6 | 548 | 150 | |
XM_006234321.2 | XP_006234383.1 | 5 - 6 | 624 | 150 | ||
XM_006234322.2 | XP_006234384.1 | 5 - 6 | 624 | 150 | ||
XM_006234323.2 | XP_006234385.1 | 5 - 6 | 624 | 150 | ||
GenBank mRNA | BC081780.1 | - | 5 - 6 | 548 | 7192454 | 150 |
X67859.1 | - | 4 - 5 | 483 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |