Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133405.2 | NP_596896.2 | 4 - 5 | 650 | 57 | |
GenBank mRNA | AY033877.1 | - | 4 - 5 | 660 | 57 | |
BC166416.1 | - | 4 - 5 | 650 | 9033484 | 57 | |
FQ211347.1 | - | 4 - 5 | 660 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |