Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234484.2 | XP_006234546.1 | 11 - 12 | 5292 | 76 | |
XM_006234485.2 | XP_006234547.1 | 11 - 12 | 5772 | 76 | ||
XM_006234486.2 | XP_006234548.1 | 11 - 12 | 5235 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |