Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108590.2 | NP_001102060.1 | 4 - 5 | 666 | 75 | |
GenBank mRNA | BC168935.1 | - | 4 - 5 | 666 | 9101840 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |