Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761999.1 | XP_008760221.1 | 3 - 4 | 496 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |