Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761944.1 | XP_008760166.1 | 9 - 10 | 1033 | 99 | |
XM_342441.7 | XP_342442.5 | 10 - 11 | 1194 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus Probe spans exons |