Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761944.1 | XP_008760166.1 | 1 - 2 | 269 | 85 | |
XM_342441.7 | XP_342442.5 | 1 - 2 | 268 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |