Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761944.1 | XP_008760166.1 | 7 - 8 | 847 | 80 | |
XM_342441.7 | XP_342442.5 | 7 - 8 | 846 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |