Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271346.1 | NP_001258275.1 | 3 - 4 | 627 | 68 | |
XM_006234498.2 | XP_006234560.1 | 3 - 4 | 620 | 68 | ||
XM_006234499.2 | XP_006234561.1 | 3 - 4 | 660 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |