Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271346.1 | NP_001258275.1 | 4 - 5 | 776 | 96 | |
XM_006234498.2 | XP_006234560.1 | 4 - 5 | 769 | 96 | ||
XM_006234499.2 | XP_006234561.1 | 4 - 5 | 809 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |