Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012220.1 | NP_001012220.1 | 1 - 2 | 345 | 66 | |
XM_006234860.2 | XP_006234922.1 | 1 - 2 | 391 | 66 | ||
GenBank mRNA | BC079422.1 | - | 1 - 2 | 345 | 7114001 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |