Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234894.2 | XP_006234956.1 | 1 - 2 | 106 | 58 | |
GenBank mRNA | FQ213399.1 | - | 1 - 2 | 45 | 58 | |
FQ231949.1 | - | 1 - 2 | 47 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |