Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134530.1 | NP_001128002.1 | 5 - 6 | 813 | 78 | |
GenBank mRNA | BC169019.1 | - | 5 - 6 | 813 | 9091657 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |