Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130422.1 | NP_569106.1 | 1 - 2 | 271 | 114 | |
XM_008765891.1 | XP_008764113.1 | 2 - 3 | 114 | |||
GenBank mRNA | AF317633.1 | - | 1 - 2 | 271 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |