Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101013.2 | NP_001094483.2 | 7 - 8 | 1223 | 82 | |
GenBank mRNA | BC168141.1 | - | 7 - 8 | 945 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |