Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053736.2 | NP_446188.2 | 2 - 3 | 366 | 94 | |
GenBank mRNA | AY029283.1 | - | 2 - 3 | 389 | 94 | |
BC104720.1 | - | 2 - 3 | 366 | 7382967 | 94 | |
FQ227402.1 | - | 2 - 3 | 403 | 94 | ||
FQ232279.1 | - | 2 - 3 | 403 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |