Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144870.1 | NP_001138342.1 | 1 - 2 | 261 | 69 | |
GenBank mRNA | BC088439.1 | - | 1 - 2 | 105 | 69 | |
DY310175.1 | - | 1 - 2 | 261 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |