Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107367.2 | NP_001100837.1 | 7 - 8 | 1071 | 128 | |
XM_008771868.1 | XP_008770090.1 | 7 - 8 | 1480 | 128 | ||
XM_008771869.1 | XP_008770091.1 | 7 - 8 | 1478 | 128 | ||
XM_008771870.1 | XP_008770092.1 | 5 - 6 | 1218 | 128 | ||
GenBank mRNA | BC162024.1 | - | 7 - 8 | 1071 | 9099401 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |