Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138547.2 | NP_612556.1 | 1 - 2 | 130 | 136 | |
GenBank mRNA | BC091123.1 | - | 1 - 2 | 130 | 7372929 | 136 |
D17310.1 | - | 1 - 2 | 318 | 136 | ||
M64393.1 | - | 1 - 2 | 129 | 136 | ||
S57790.1 | - | 1 - 2 | 88 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |