Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106125.1 | NP_001099595.1 | 2 - 3 | 118 | 71 | |
XM_006254307.2 | XP_006254369.1 | 3 - 4 | 4032 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |