Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104630.1 | NP_001098100.1 | 6 - 7 | 1011 | 154 | |
XM_006254266.2 | XP_006254328.1 | 6 - 7 | 2638 | 154 | ||
GenBank mRNA | AY724491.1 | - | 6 - 7 | 1011 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |