Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031641.1 | NP_001026811.1 | 1 - 2 | 135 | 67 | |
GenBank mRNA | BC098718.1 | - | 1 - 2 | 135 | 7366394 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |