Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044247.1 | NP_001037712.1 | 3 - 4 | 450 | 71 | |
XM_006253100.2 | XP_006253162.1 | 3 - 4 | 1880 | 71 | ||
XM_008771246.1 | XP_008769468.1 | 3 - 4 | 883 | 71 | ||
XM_008771247.1 | XP_008769469.1 | 3 - 4 | 478 | 71 | ||
XM_008771248.1 | XP_008769470.1 | 3 - 4 | 1723 | 71 | ||
GenBank mRNA | BC098935.1 | - | 3 - 4 | 450 | 7445963 | 71 |
FQ214804.1 | - | 3 - 4 | 400 | 71 | ||
FQ215658.1 | - | 3 - 4 | 322 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |