Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047086.1 | NP_001040551.1 | 7 - 8 | 813 | 76 | |
XM_006253139.2 | XP_006253201.1 | 7 - 8 | 1074 | 76 | ||
XM_008771256.1 | XP_008769478.1 | 7 - 8 | 909 | 76 | ||
GenBank mRNA | BC166593.1 | - | 7 - 8 | 854 | 7934031 | 76 |
DQ674266.1 | - | 7 - 8 | 813 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |