Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134686.2 | NP_001128158.1 | 2 - 3 | 555 | 77 | |
NM_022619.3 | NP_072141.2 | 2 - 3 | 555 | 77 | ||
XM_008771281.1 | XP_008769503.1 | 2 - 3 | 5634 | 77 | ||
XM_008771282.1 | XP_008769504.1 | 3 - 4 | 514 | 77 | ||
XM_008771283.1 | XP_008769505.1 | 2 - 3 | 5634 | 77 | ||
GenBank mRNA | AF245001.1 | - | 2 - 3 | 556 | 77 | |
AF245002.1 | - | 2 - 3 | 556 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |