Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251938.2 | XP_006252000.1 | 3 - 4 | 7401 | 63 | |
XM_006251939.2 | XP_006252001.1 | 3 - 4 | 7371 | 63 | ||
XM_008770674.1 | XP_008768896.1 | 4 - 5 | 7335 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |