Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106034.2 | NP_001099504.2 | 4 - 5 | 794 | 135 | |
GenBank mRNA | CX569344.1 | - | 3 - 4 | 310 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |