Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017257.2 | NP_058953.1 | 5 - 6 | 263 | 71 | |
GenBank mRNA | BC058486.1 | - | 5 - 6 | 301 | 6918985 | 71 |
BC097331.1 | - | 5 - 6 | 263 | 7389180 | 71 | |
D45250.1 | - | 5 - 6 | 313 | 71 | ||
FQ226805.1 | - | 5 - 6 | 294 | 71 | ||
FQ227049.1 | - | 5 - 6 | 307 | 71 | ||
FQ232840.1 | - | 5 - 6 | 293 | 71 | ||
FQ233763.1 | - | 5 - 6 | 289 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |