Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001078405.6 | XP_001078405.4 | 7 - 8 | 673 | 69 | |
XM_008770715.1 | XP_008768937.1 | 7 - 8 | 689 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |