Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098803.1 | NP_001092273.1 | 4 - 5 | 3157 | 59 | |
XM_008770677.1 | XP_008768899.1 | 3 - 4 | 4932 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |