Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031056.1 | NP_112318.1 | 4 - 5 | 852 | 70 | |
GenBank mRNA | BC072509.1 | - | 4 - 5 | 883 | 7103279 | 70 |
X83537.1 | - | 4 - 5 | 861 | 70 | ||
X91785.1 | - | 4 - 5 | 852 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |