Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030850.1 | NP_110477.1 | 4 - 5 | 545 | 80 | |
GenBank mRNA | AF038870.1 | - | 4 - 5 | 545 | 80 | |
BC078811.1 | - | 4 - 5 | 532 | 7130736 | 80 | |
U96133.1 | - | 4 - 5 | 534 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |