Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108541.1 | NP_001102011.1 | 11 - 12 | 1030 | 99 | |
XM_006231815.2 | XP_006231877.1 | 12 - 13 | 1276 | 99 | ||
GenBank mRNA | BC166792.1 | - | 12 - 13 | 1202 | 7937195 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |