Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008372.1 | NP_001008373.1 | 12 - 13 | 1542 | 111 | |
XM_006231776.2 | XP_006231838.1 | 12 - 13 | 1532 | 111 | ||
XM_006231778.2 | XP_006231840.1 | 12 - 13 | 1569 | 111 | ||
XM_008760639.1 | XP_008758861.1 | 13 - 14 | 1645 | 111 | ||
GenBank mRNA | BC085771.1 | - | 12 - 13 | 1542 | 7191068 | 111 |
FQ211725.1 | - | 12 - 13 | 1561 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |