Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022509.1 | NP_071954.1 | 3 - 4 | 309 | 65 | |
GenBank mRNA | AF044910.1 | - | 3 - 4 | 289 | 65 | |
AY876898.1 | - | 3 - 4 | 279 | 65 | ||
BC062404.1 | - | 3 - 4 | 294 | 5621519 | 65 | |
FQ212707.1 | - | 3 - 4 | 305 | 65 | ||
U75369.1 | - | 3 - 4 | 309 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |