Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106406.1 | NP_001099876.1 | 8 - 9 | 1665 | 142 | |
XM_006231887.2 | XP_006231949.1 | 8 - 9 | 1442 | 142 | ||
XM_008760666.1 | XP_008758888.1 | 10 - 11 | 1437 | 142 | ||
GenBank mRNA | BC078983.1 | - | 8 - 9 | 1882 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |